Learn More
Invitrogen™ FNBP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589450
Description
Immunogen sequence: MGKKSRAVPG RRPILQLSPP GPRGSTPGRD PEPEPDTEPD STAAVPSQPA PSAATTTTTA VTAAAASDDS PSEDEQEAVQ EVPRVVQNPP KPVMTTRPTA VKATGGLCLL GAYADSDDDD NDVSEKLAQS KETNGNQSTD IDSTLANFLA EIDAITAPQP AAPVGASAPP PTPPRPEPKE AATSTLSSST SNGTDSTQTS GWQYDTQCSL AGVGIEMGDW QEVWDENTGC YYYWNTQTNE VTWELPQYLA TQVQGLQHYQ; Positive Samples: Mouse kidney.
FNBP4 encodes a protein containing two tryptophan-rich WW domains that binds the proline-rich formin homology 1 domains of formin family proteins, suggesting a role in the regulation of cytoskeletal dynamics during cell division and migration. It also binds intersectin family proteins suggesting a role in the maintenance of membrane curvature at sites of nascent vesicle formation. Naturally occurring mutations in this gene are associated with Waardenburg anophthalmia syndrome.
Specifications
| FNBP4 | |
| Polyclonal | |
| Unconjugated | |
| FNBP4 | |
| FBP30; Fnbp30; Fnbp4; formin binding protein 30; formin binding protein 4; formin-binding protein 30; Formin-binding protein 4; Kiaa1014; mKIAA1014 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55935 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6ZQ03 | |
| FNBP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human FNBP4 (NP_0561232). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.