missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FNBP1L Polyclonal antibody specifically detects FNBP1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | FNBP1L |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1orf39, chromosome 1 open reading frame 39, FLJ20275, formin binding protein 1-like, formin-binding protein 1-like, toca-1, TOCA1Transducer of Cdc42-dependent actin assembly protein 1, transducer of Cdc42-dependent actin assembly 1 |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 450 and 500 of human Transducer of Cdc42-Dependent Actin Assembly Protein 1 using the numbering given in entry NP_001157945.1 (GeneID 54874). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?