missing translation for 'onlineSavingsMsg'
Learn More

FNBP1L Antibody, Biotin, Novus Biologicals™

Product Code. 18430031 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430031 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430031 Supplier Novus Biologicals Supplier No. NBP180569B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FNBP1L Polyclonal antibody specifically detects FNBP1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FNBP1L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias C1orf39, chromosome 1 open reading frame 39, FLJ20275, formin binding protein 1-like, formin-binding protein 1-like, toca-1, TOCA1Transducer of Cdc42-dependent actin assembly protein 1, transducer of Cdc42-dependent actin assembly 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 450 and 500 of human Transducer of Cdc42-Dependent Actin Assembly Protein 1 using the numbering given in entry NP_001157945.1 (GeneID 54874).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54874
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.