missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FN3K Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570339
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%.
FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.
Specifications
| FN3K | |
| Polyclonal | |
| Unconjugated | |
| Fn3k | |
| 2310074G21Rik; FN3K; Fnsk; fructosamine 3 kinase; fructosamine 3-kinase; fructosamine-3-kinase; Protein-psicosamine 3-kinase FN3K; Protein-ribulosamine 3-kinase FN3K | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 498034 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Fn3k | |
| synthetic peptide directed towards the N-terminal region of rat FN3K. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction