missing translation for 'onlineSavingsMsg'
Learn More

FN3K Antibody (4F2), Novus Biologicals™

Product Code. 18421969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421969 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421969 Supplier Novus Biologicals Supplier No. H00064122M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FN3K Monoclonal antibody specifically detects FN3K in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FN3K
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4F2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_071441
Gene Alias EC 2.7.1.-, fructosamine 3 kinase, fructosamine-3-kinase
Host Species Mouse
Immunogen FN3K (NP_071441.1, 61 a.a. ∽ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ALRSTGLVRVPRPMKVIDLPGGGAAFVMEHLKMKSLSSQASKLGEQMADLHLYNQKLREKLKEEENTVGRRGEGAEPQYVDKFGFHTVTCCGFIPQVNEWQDDWPTFFAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 64122
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.