missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMRP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £409.00
Specifications
| Antigen | FMRP |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684745
|
Novus Biologicals
NBP2-38783-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18107488
|
Novus Biologicals
NBP2-38783 |
0.1 mL |
£409.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMRP Polyclonal specifically detects FMRP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FMRP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q06787 | |
| 2332 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RIEAENEKNVPQEEEIMPPNSLPSNNSRVGPNAPEEKKHLDIKENSTHFSQPNSTKV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Epigenetics, microRNA | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FMRPFRAXAPOF, fragile X mental retardation 1, fragile X mental retardation 1 protein, MGC87458, POF1, premature ovarian failure 1, Protein FMR-1 | |
| FMR1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title