missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-valine, TRC

Product Code. 30389023
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30389023 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389023 Supplier Toronto Research Chemicals Supplier No. TRCF68000010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-valine
CAS 68858-20-8
Molecular Formula C20H21NO4
Quantity 10 g
Synonym N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-valine, (2S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-methylbutanoic Acid, (9-Fluorenylmethoxycarbonyl)valine, (S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoic Acid, (S)-N-Fmoc-valine, 125: PN: WO2004099237 PAGE: 36 claimed protein, 221: PN: US20070042401 PAGE: 29 claimed protein, 912: PN: WO2006135786 PAGE: 59 claimed protein, FMOC-L-valine, Fmoc-L-Val-OH, N-(9-Fluorenylmethoxycarbonyl)-L-valine, N-(9-Fluorenylmethoxycarbonyl)valine, NSC 334304
InChI Formula InChI=1S/C20H21NO4/c1-12(2)18(19(22)23)21-20(24)25-11-17-15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10,12,17-18H,11H2,1-2H3,(H,21,24)(H,22,23)/t18-/m0/s1
SMILES CC(C)[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoic acid
Molecular Weight (g/mol) 339.39
Formula Weight 339.1471
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.