missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-n-me-lys(boc)-oh, TRC

Product Code. 30400911
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400911 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400911 Supplier Toronto Research Chemicals Supplier No. TRCF64849825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-n-me-lys(boc)-oh
CAS 197632-76-1
Molecular Formula C27 H34 N2 O6
Quantity 25 mg
InChI Formula InChI=1S/C27H34N2O6/c1-27(2,3)35-25(32)28-16-10-9-15-23(24(30)31)29(4)26(33)34-17-22-20-13-7-5-11-18(20)19-12-6-8-14-21(19)22/h5-8,11-14,22-23H,9-10,15-17H2,1-4H3,(H,28,32)(H,30,31)/t23-/m0/s1
SMILES CN([C@@H](CCCCNC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2ccccc2c3ccccc13
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Molecular Weight (g/mol) 482.569
Formula Weight 482.242
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.