missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-N-Me-Ala-OH, MedChemExpress

Product Code. 16499326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499326 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499326 Supplier Medchem Express Supplier No. HYW008235
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fmoc-N-Me-Ala-OH, an N-Fmoc-N-methyl amino acid, is available for the peptide-coupling reaction.

Fmoc-N-Me-Ala-OH, an N-Fmoc-N-methyl amino acid, is available for the peptide-coupling reaction.

Specifications

Chemical Name or Material Fmoc-N-Me-Ala-OH
CAS 84000-07-7
Color White
Purity Grade Notes Research
Molecular Formula C19H19NO4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (307.35 mM; Need ultrasonic)
SMILES C[C@H](N(C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)C)C(O)=O
Molecular Weight (g/mol) 325.36
Formula Weight 325.36
Percent Purity 97.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.