missing translation for 'onlineSavingsMsg'
Learn More

FMOC-L-Norleucine, 98%, ACROS Organics™

Brand:  Acros Organics 442720050

Code : 29

Additional Details : CAS Number : 77284-32-3 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11975571

  • £61.50 / 5g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

FMOC-L-Norleucine
98%
353.42
≥97.5%
C21H23NO4
(S)-2-(FMOC-amino)hexanoic acid, FMOC-Nle-OH

Specifications

FMOC-L-Norleucine
137° to 145°C
77284-32-3
-18.0° to -20.0° (20°C, 589nm) (c-1, DMF)
≥97.5%
C21H23NO4
(S)-2-(FMOC-amino)hexanoic acid, FMOC-Nle-OH
White to off-white
5g
Authentic
Crystalline Powder
98%
353.42
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FMOC-L-Norleucine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS