missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Fmoc-L-Isoleucine, 10/Package
Click to view available options
Quantity:
10 Ea.
Unit Size:
Each
Description
- Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Content And Storage | Each package contains 10 cartridges. |
| Format | Cartridge |
| For Use With (Equipment) | 433A Peptide Synthesizer |
| For Use With (Application) | Protein Sequencing & Peptide Synthesis |
| Product Type | Peptide Synthesis Reagents |
| Quantity | 10 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction