missing translation for 'onlineSavingsMsg'
Learn More

FMOC-L-α-Alanine monohydrate, 95%, Thermo Scientific™

Product Code. 10017899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10017899 5 g 5g
10381140 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10017899 Supplier Thermo Scientific Acros Supplier No. 437610050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 207291-76-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
InChI Key GAPWKFLOMOFHGO-UHFFFAOYNA-N
Synonym n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate
PubChem CID 18528118
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate
SMILES O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 146.8°C to 148.1°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation -20
Formula Weight 329.36
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material FMOC-L-α-Alanine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.