missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-l-2-pyridylalanine, TRC

Product Code. 30392318
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392318 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392318 Supplier Toronto Research Chemicals Supplier No. TRCF8610331G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-L-2-Pyridylalanine
CAS 185379-40-2
Molecular Formula C23 H20 N2 O4
Quantity 1 g
InChI Formula InChI=1S/C23H20N2O4/c26-22(27)21(13-15-7-5-6-12-24-15)25-23(28)29-14-20-18-10-3-1-8-16(18)17-9-2-4-11-19(17)20/h1-12,20-21H,13-14H2,(H,25,28)(H,26,27)/t21-/m0/s1
SMILES OC(=O)[C@H](Cc1ccccn1)NC(=O)OCC2c3ccccc3c4ccccc24
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-pyridyl)propanoic acid
Molecular Weight (g/mol) 388.42
Formula Weight 388.1423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.