missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-L-2-furylalanine, TRC

Product Code. 30393783
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393783 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393783 Supplier Toronto Research Chemicals Supplier No. TRCF6797281G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-L-2-furylalanine
CAS 159611-02-6
Molecular Formula C22 H19 N O5
Quantity 1 g
InChI Formula InChI=1S/C22H19NO5/c24-21(25)20(12-14-6-5-11-27-14)23-22(26)28-13-19-17-9-3-1-7-15(17)16-8-2-4-10-18(16)19/h1-11,19-20H,12-13H2,(H,23,26)(H,24,25)/t20-/m0/s1
SMILES OC(=O)[C@H](Cc1occc1)NC(=O)OCC2c3ccccc3c4ccccc24
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-furyl)propanoic acid
Molecular Weight (g/mol) 377.39
Formula Weight 377.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.