missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-Glycinol, TRC

Product Code. 30391776
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391776 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391776 Supplier Toronto Research Chemicals Supplier No. TRCF6199755G
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fmoc-Glycinol
CAS 105496-31-9
Molecular Formula C17H17NO3
Quantity 5 g
Synonym N-(2-Hydroxyethyl)carbamic Acid 9H-Fluoren-9-ylmethyl Ester, (2-Hydroxyethyl)carbamic Acid 9H-Fluoren-9-ylmethyl Ester, Fmoc-2-aminoethanol, N-(9-Fluorenylmethoxycarbonyl)-2-aminoethanol, N-Fmoc-2-aminoethanol
InChI Formula InChI=1S/C17H17NO3/c19-10-9-18-17(20)21-11-16-14-7-3-1-5-12(14)13-6-2-4-8-15(13)16/h1-8,16,19H,9-11H2,(H,18,20)
SMILES OCCNC(=O)OCC1c2ccccc2c3ccccc13
IUPAC Name 9H-fluoren-9-ylmethyl N-(2-hydroxyethyl)carbamate
Molecular Weight (g/mol) 283.32
Formula Weight 283.1208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.