missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-D-Gln(Trt)-OH, TRC

Product Code. 30179612
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179612 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179612 Supplier Toronto Research Chemicals Supplier No. TRCF680028100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 200623-62-7
Molecular Formula C35 H28 N4 O5
Synonym N2-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-(triphenylmethyl)-D-glutamine;
InChI Formula InChI=1S/C35H28N4O5/c1-35-31(43-3)23(37(2)34(42)18-11-5-4-6-12-18)17-24(44-35)38-21-15-9-7-13-19(21)25-27-28(33(41)36-32(27)40)26-20-14-8-10-16-22(20)39(35)30(26)29(25)38/h4-16,23-24,31H,17H2,1-3H3,(H,36,40,41)/t23-,24+,31-,35-/m0/s1
SMILES CO[C@H]1[C@H](C[C@H]2O[C@]1(C)n3c4ccccc4c5c6C(=O)NC(=O)c6c7c8ccccc8n2c7c35)N(C)C(=O)c9ccccc9
IUPAC Name N-[(2S,3S,4S,6R)-3-methoxy-2-methyl-16,18-dioxo-29-oxa-1,7,17-triazaoctacyclo[12.12.2.12,6.07,28.08,13.015,19.020,27.021,26]nonacosa-8(13),9,11,14,19,21(26),22,24,27-nonaen-4-yl]-N-methylbenzamide
Molecular Weight (g/mol) 584.621
Quantity 100 mg
Formula Weight 584.206
Shipping Condition Room Temperature
Chemical Name or Material Fmoc-D-Gln(Trt)-OH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.