missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-1,6-diaminohexane, MedChemExpress

Product Code. 16480976
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480976 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16480976 Supplier Medchem Express Supplier No. HY103664
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fmoc-1,6-diaminohexane is an analog of Osw-1 which has the potential for Alzheimer's disease and cancer treatment from patent US 20140135279 A1.

Fmoc-1,6-diaminohexane is an analog of Osw-1 which has the potential for Alzheimer's disease and cancer treatment from patent US 20140135279 A1.

Specifications

Chemical Name or Material Fmoc-1,6-diaminohexane
CAS 166410-37-3
Color White
Purity Grade Notes Research
Molecular Formula C21H26N2O2
Quantity 200 mg
Solubility Information DMSO : 150 mg/mL (443.21 mM; Need ultrasonic and warming)
SMILES O=C(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)NCCCCCCN
Molecular Weight (g/mol) 338.44
Formula Weight 338.44
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.