missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569370
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 85%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%.
FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Specifications
| FMO4 | |
| Polyclonal | |
| Unconjugated | |
| Fmo4 | |
| D1Ertd532e; Dimethylaniline monooxygenase [N-oxide-forming] 4; dimethylaniline oxidase 4; flavin containing dimethylaniline monoxygenase 4; flavin containing monooxygenase 4; flavin-containing monooxygenase 4; flavin-containing monooxygenase FMO4; FMO 1E1; FMO 4; FMO2; Fmo4; Hepatic flavin-containing monooxygenase 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2329 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P31512 | |
| Fmo4 | |
| synthetic peptide directed towards the N terminal region of human FMO4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction