missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33583
This item is not returnable.
View return policy
Description
FMO3 Polyclonal specifically detects FMO3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FMO3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FMO3 | |
| Recombinant protein encompassing a sequence within the center region of human FMO3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2328 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.91mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU | |
| Rabbit | |
| 60 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction