missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16861-1-AP-150UL
This item is not returnable.
View return policy
Description
The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.Specifications
| FMO2 | |
| Polyclonal | |
| Unconjugated | |
| FMO2 | |
| Dimethylaniline oxidase 2, FMO 1B1, FMO 2, FMO1B1, FMO2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2327, 246245, 55990 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6IRI9, Q8K2I3, Q99518 | |
| Fmo2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction