missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FMNL3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA523317

Product Code. 13277898

  • £481.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence homology: Chimpanzee (100%), Bovine (72%).

FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FMNL3
Polyclonal
Unconjugated
FMNL3
2700073B04Rik; FBP11; FHOD3; FMNL3; Formin homology 2 domain-containing protein 3; formin like 3; formin-like 3; formin-like 3 protein; formin-like protein 3; Frl2; KIAA2014; mKIAA2014; WBP3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3
Rabbit
Protein G
RUO
100059687, 22379, 300225, 486555, 712659, 91010
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q6ZPF4, Q8IVF7
FMNL3
A portion of amino acids 150-200 of human Formin-like protein 3.
100 μg
Primary
Human, Mouse, Rat, Canine, Monkey, Horse, Rhesus Macaque
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.