missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMNL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554720
This item is not returnable.
View return policy
Description
Immunogen sequence: LDLENENMMR VAELEKQLLQ REKELESIKE TYENTSHQVH TLRRLIKEKE EAFQRRCHLE PNVRGLESVD SEALARVGPA ELSEGMPPSD LDLLAPAPPP EEVLPLPPPP APPLPPPPPP LPDKCP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
FMNL3, also known as Formin-like protein 3, is a 1028 amino acid containing protein involved in immunity or defense. This developmental protein is implicated in polarity control, invasion, migration, or metastasis through regulation of the Rho-related signaling pathway. It belongs to the formin homology family and contains a FH2 (formin homology 2) domain and a GBD/FH3 (Rho GTPase-binding/formin homology 3) domain and has high sequence identity to the mouse Wbp3 protein. Two different isoforms have been described. Itis suspectedbe involved in gastric cancer.
Specifications
| FMNL3 | |
| Polyclonal | |
| Unconjugated | |
| FMNL3 | |
| 2700073B04Rik; FBP11; FHOD3; FMNL3; Formin homology 2 domain-containing protein 3; formin like 3; formin-like 3; formin-like 3 protein; formin-like protein 3; Frl2; KIAA2014; mKIAA2014; WBP3; WBP-3; WW domain binding protein 3; WW domain-binding protein 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 91010 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVF7 | |
| FMNL3 | |
| Recombinant protein corresponding to Human FMNL3. Recombinant protein control fragment (Product #RP-102029). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction