Learn More
FMNL2 Mouse, anti-Human, Clone: 3E8C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68551-1-IG-20UL
This item is not returnable.
View return policy
Description
The FMNL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FMNL2. This antibody recognizes human antigen. The FMNL2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FMNL2 | |
| 3E8C12 | |
| FHOD2, FLJ37546, FMNL2, formin like 2, Formin like protein 2, KIAA1902 | |
| 20 μL | |
| 114793 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |