missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMNL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534477
This item is not returnable.
View return policy
Description
A suggested positive control is EL4 cell lysate. PA5-34477 can be used with blocking peptide PEP-1519.
The Formin-like protein 1 (FRL1) gene encodes a formin-related protein which has been implicated in morphogenesis, cyokinesis, and cell polarity. Formins are a conserved class of proteins expressed in all eukaryotes and have one DAD (diaphanous autoregulatory domain), one FH2 (formin homology 2) domain and one GBD/FH3 (Rho GTPase-binding / formin homology 3) domain. FRL1 is located in the cytoplasm and is highly expressed in the spleen, lymph node and bone marrow cells. FRL1 possibly has a role in the control of cell motility, survival of macrophages and cytoskeletal organization.
Specifications
| FMNL1 | |
| Polyclonal | |
| Unconjugated | |
| FMNL1 | |
| 8030453N10Rik; AI553564; C17orf1; C17orf1B; CLL-associated antigen KW-13; FHOD4; Fmnl; Fmnl1; Fnrl; formin like 1; formin-like 1; formin-like protein 1; formin-related gene in leukocytes; formin-related protein; Frl; FRL1; Frls; KW-13; Leukocyte formin; lymphocyte specific formin related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 287746, 57778, 752 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95466, Q9JL26 | |
| FMNL1 | |
| A 16 amino acid peptide near the carboxy terminus of human FRL1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction