missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FMNL1 Polyclonal antibody specifically detects FMNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | FMNL1 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C17orf1BMGC133052, C17orf1KW-13, CLL-associated antigen KW-13, FHOD4, FMNLMGC1894, formin-like, formin-like 1, formin-like protein 1, Leukocyte formin, MGC21878 |
| Host Species | Rabbit |
| Immunogen | Rabbit polyclonal FMNL1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human FMNL1. The immunogen is located within the last 50 amino acids of FMNL1. Amino Acid Squence: CTVPFTARTGKRTSRLL |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?