missing translation for 'onlineSavingsMsg'
Learn More

FMNL1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18405047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405047 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405047 Supplier Novus Biologicals Supplier No. NBP281988AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FMNL1 Polyclonal antibody specifically detects FMNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMNL1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias C17orf1BMGC133052, C17orf1KW-13, CLL-associated antigen KW-13, FHOD4, FMNLMGC1894, formin-like, formin-like 1, formin-like protein 1, Leukocyte formin, MGC21878
Host Species Rabbit
Immunogen Rabbit polyclonal FMNL1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human FMNL1. The immunogen is located within the last 50 amino acids of FMNL1. Amino Acid Squence: CTVPFTARTGKRTSRLL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.