missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FMF-04-159-2
GREENER_CHOICE

Product Code. 18797136
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18797136

Brand: Tocris Bioscience™ 7158/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent covalent CDK14 and CDK16 inhibitor; also inhibits other TAIRE kinase family members

FMF-04-159-2 is a potent inhibitor of CDK14 and CDK16 (IC50 = 40 nM for CDK14 in cellular BRET assay; IC50 values are 88 and 10 nM for CDK14 and CDK16 in kinase activity inhibition assay, respectively). Inhibits other TAIRE kinase family members at 1 μM (CDK17 and CDK18). Also binds CDK2 (IC50 = 256 nM). Displays covalent binding of CDK14, with binding sustained after washout. Causes cell cycle arrest at G2/M in cancer cell lines. Reversible control FMF-04-159-R (Cat. No. 7159) also available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[1-[[3-[[(2 E)-4-(Dimethylamino)-1-oxo-2-buten-1-yl]amino]phenyl]sulfonyl]-4-piperidinyl]-4-[(2,4,6- trichlorobenzoyl)amino]-1 H-pyrazole-3-carboxamide
CAS 2364489-81-4
Quantity 10 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C28H30Cl3N7O5S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.