missing translation for 'onlineSavingsMsg'
Learn More

FMC63 scFv Mouse, PE, Clone: Y45, AcroBiosystems

Mouse Monoclonal Antibody

£374.00 - £2280.00

Specifications

Antigen FMC63 scFv
Clone Y45
Classification Monoclonal
Conjugate PE
Host Species Human (HEK293)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17101491
View Documents
ACROBiosystems
FM3-HPY53-25TESTS
25 Tests
£374.00
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17146321
View Documents
ACROBiosystems
FM3-HPY53-200tests
200 Tests
£2280.00
200 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.
Specifications

Specifications

FMC63 scFv
Monoclonal
Human (HEK293)
IgG1
-20°C, aviod light
Y45
PE
RUO
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.