Learn More
FMC63 scFv Mouse, FITC, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Brand: ACROBiosystems FM3-FY45-200tests
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Specifications
| FMC63 scFv | |
| Y45 | |
| Hybridoma | |
| RUO | |
| -20°C, aviod light |
| Monoclonal | |
| FITC | |
| 200 Tests | |
| Primary | |
| IgG1 |