missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FM19G11

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
FM19G11 is a hypoxia inducible factor (HIF) α-subunit inhibitor (IC50 = 80 nM in hypoxia induced luciferase assay). Inhibits transcriptional activity of HIFα isoforms. Directly regulates expression and inhibits transcriptional activity of pluripotency markers (Sox2 and Oct4) in rodent and human stem cells under hypoxic conditions, driving differentiation. Improves locomotion after severe spinal cord injury in mice, alone or in combination with transplantation of ependymal stem cells of the spinal cord. Also inhibits O6-methylguanine DNA methyltransferase (MGMT) via HIF1α pathway, under normal and hypoxic conditions in glioblastoma cell lines.
Specifications
Specifications
| Chemical Name or Material | 2-Oxo-2-(p-tolyl)ethyl 3-(2,4-dinitrobenzamido)benzoate |
| CAS | 329932-55-0 |
| Quantity | 25 mg |
| Molecular Formula | C23H17N3O8 |
| Formula Weight | 463.4 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction