missing translation for 'onlineSavingsMsg'
Learn More

FM-381, MedChemExpress

Product Code. 16453698
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453698 10 mg 10mg
16450087 50 mg 50mg
16446437 5 mg 5mg
16444776 100 mg 100mg
16422836 25 mg 25mg
16410076 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16453698 Supplier Medchem Express Supplier No. HY102046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FM-381 is a potent covalent reversible inhibitor of JAK3 targeting the unique Cys909. FM-381 has an IC50 of 127 pM for JAK3, with 410, 2700 and 3600-fold selectivity over JAK1, JAK2 and TYK2, respectively.

FM-381 is a potent covalent reversible inhibitor of JAK3 targeting the unique Cys909. FM-381 has an IC50 of 127 pM for JAK3, with 410, 2700 and 3600-fold selectivity over JAK1, JAK2 and TYK2, respectively.

Specifications

Chemical Name or Material FM-381
CAS 2226521-65-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H24N6O2
Quantity 10 mg
Solubility Information DMSO : 8.33 mg/mL (19.44 mM; Need ultrasonic)
SMILES CN(C(/C(C#N)=C/C1=CC=C(C2=NC3=CN=C(NC=C4)C4=C3N2C5CCCCC5)O1)=O)C
Molecular Weight (g/mol) 428.49
Formula Weight 428.49
Percent Purity 98.25%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.