missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLYWCH2 Antibody (OTI3G3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72341MFV610
This item is not returnable.
View return policy
Description
FLYWCH2 Monoclonal antibody specifically detects FLYWCH2 in Human samples. It is validated for Flow Cytometry
Specifications
| FLYWCH2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI3G3 | |
| Flow Cytometry | |
| FLYWCH family member 2 | |
| Full length human recombinant protein of human FLYWCH2 (NP_612448) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology | |
| 114984 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction