missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLVCR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26841-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the major facilitator superfamily of transporter proteins. The encoded protein is a heme transporter that may play a critical role in erythropoiesis by protecting developing erythroid cells from heme toxicity. This gene may play a role in posterior column ataxia with retinitis pigmentosa and the hematological disorder Diamond-Blackfan syndrome.Specifications
| FLVCR1 | |
| Polyclonal | |
| Unconjugated | |
| FLVCR1 | |
| FLVCR, FLVCR1, hFLVCR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 226844, 28982 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RXV4, Q9Y5Y0 | |
| FLVCR1 | |
| FLVCR1 Fusion Protein Ag25220 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction