missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Fluvoxamine maleate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective serotonin reuptake inhibitor; antidepressant. Binds to the human 5-HT transporter with a Ki of 1.6 nmol/l.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 434.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 434.41 |
| Chemical Name or Material | Fluvoxamine maleate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction