missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxamine Maleate, Mikromol™

Product Code. 17911206
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17911206

Brand: Mikromol MM0058.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fluvoxamine Maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluvoxamine maleate
CAS 61718-82-9
Analyte or Component Names Fluvoxamine maleate
Shipping Condition Room Temperature
Molecular Formula C15 H21 F3 N2 O2 . C4 H4 O4
Certifications/Compliance ISO 17034
Synonym Fluvoxamine maleate, 2-[[[(1E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethanamine (Z)-butenedioate, 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (1E)-, (2Z)-2-butenedioate (1:1), 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, (Z)-2-butenedioate (1:1), Avoksin, Avoxin, DU 23000, Depromel, Dumirox, Faverin, Faverin 50, Faverine, Fevarin, Floxyfral, Luvox, MK 264, Maveral, NSC 309469
InChI Formula InChI=1S/C15H21F3N2O2.C4H4O4/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18;5-3(6)1-2-4(7)8/h5-8H,2-4,9-11,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b20-14+;2-1-
SMILES COCCCC\C(=N/OCCN)\c1ccc(cc1)C(F)(F)F.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethanamine
Molecular Weight (g/mol) 434.41
Quantity 250 mg
Formula Weight 434.1665 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.