missing translation for 'onlineSavingsMsg'
Learn More

SR-3306, MedChemExpress

Product Code. 16448097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16448097

Brand: Medchem Express HY12829

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR-3306 is a selective, potent, highly brain penetrant JNK inhibitor.

SR-3306 is a selective, potent, highly brain penetrant JNK inhibitor.

Specifications

Chemical Name or Material SR-3306
CAS 1128096-91-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H26N8O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (254.81 mM; Need ultrasonic)
SMILES CC(C=C1)=NC=C1C2=NN(C=N2)C3=CC=C(NC4=NC=CC(C5=CC=CC(N6CCOCC6)=C5)=N4)C=C3
Molecular Weight (g/mol) 490.56
Formula Weight 490.56
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.