missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxamine maleate, MedChemExpress

Product Code. 16408348
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408348

Brand: Medchem Express HYB0103A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fluvoxamine maleate (DU-23000 maleate) is an antidepressant which functions pharmacologically as a selective serotonin reuptake inhibitor.

Fluvoxamine maleate (DU-23000 maleate) is an antidepressant which functions pharmacologically as a selective serotonin reuptake inhibitor.

Specifications

Chemical Name or Material Fluvoxamine maleate
CAS 61718-82-9
Color White
Purity Grade Notes Research
Molecular Formula C19H25F3N2O6
Quantity 10 mg
Synonym DU-23000 maleate
Solubility Information DMSO : ≥ 100 mg/mL (230.20 mM) ∣H2O : 20 mg/mL (46.04 mM; Need ultrasonic)
SMILES O=C(O)/C=C\C(O)=O.FC(C1=CC=C(/C(CCCCOC)=N/OCCN)C=C1)(F)F
Molecular Weight (g/mol) 434.41
Formula Weight 434.41
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.