missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin sodium, MedChemExpress

Product Code. 16412267
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16412267

Brand: Medchem Express HY14664A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. Fluvastatin sodium protects vascular smooth muscle cells against oxidative stress through the Nrf2-dependent antioxidant pathway.

Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. Fluvastatin sodium protects vascular smooth muscle cells against oxidative stress through the Nrf2-dependent antioxidant pathway.

Specifications

Chemical Name or Material Fluvastatin sodium
CAS 93957-55-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H25FNNaO4
Quantity 50 mg
Synonym XU 62-320
Solubility Information H2O : 50 mg/mL (115.35 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (115.35 mM; Need ultrasonic)
SMILES O=C(O[Na])C[C@H](O)C[C@H](O)/C=C/C(N1C(C)C)=C(C2=CC=C(F)C=C2)C3=C1C=CC=C3
Molecular Weight (g/mol) 433.45
Formula Weight 433.45
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.