missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin N-Ethyl Sodium Salt (Fluvastatin Impurity), TRC

Product Code. 30389932
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389932 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389932 Supplier Toronto Research Chemicals Supplier No. TRCF60124010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluvastatin N-Ethyl Sodium Salt (Fluvastatin Impurity)
CAS 93936-64-2
Molecular Formula C23H23FNNaO4
Quantity 10 mg
Synonym 6-Heptenoic acid, 7-[1-ethyl-3-(4-fluorophenyl)-1H-indol-2-yl]-3,5-dihydroxy-, sodium salt (1:1), (3R,5S,6E)-rel-, 6-Heptenoic acid, 7-[1-ethyl-3-(4-fluorophenyl)-1H-indol-2-yl]-3,5-dihydroxy-, monosodium salt, [R*,S*-(E)]- (9CI), 6-Heptenoic acid, 7-[1-ethyl-3-(4-fluorophenyl)-1H-indol-2-yl]-3,5-dihydroxy-, monosodium salt, [R*,S*-(E)]-(±)-
InChI Formula InChI=1S/C23H24FNO4.Na/c1-2-25-20-6-4-3-5-19(20)23(15-7-9-16(24)10-8-15)21(25)12-11-17(26)13-18(27)14-22(28)29;/h3-12,17-18,26-27H,2,13-14H2,1H3,(H,28,29);/q;+1/p-1/b12-11+;/t17-,18-;/m1./s1
SMILES [Na+].CCn1c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c(c2ccc(F)cc2)c3ccccc13
IUPAC Name sodium;(E,3R,5S)-7-[1-ethyl-3-(4-fluorophenyl)indol-2-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol) 419.42
Formula Weight 419.1509
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.