missing translation for 'onlineSavingsMsg'
Learn More

Fluvastatin 3-Oxo-4,6-diene O-tert-Butyl Ester, TRC

Product Code. 30400775
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400775 50 mg
30400085 10 mg
30400912 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400775 Supplier Toronto Research Chemicals Supplier No. TRCF60130050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920275-08-7
Molecular Formula C28 H30 F N O3
Molecular Weight (g/mol) 447.541
Synonym (4E,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-oxo-4,6-heptadienoic Acid 1,1-Dimethylethyl Ester
IUPAC Name tert-butyl (4E,6E)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3-oxohepta-4,6-dienoate
SMILES CC(C)n1c(\C=C\C=C\C(=O)CC(=O)OC(C)(C)C)c(c2ccc(F)cc2)c3ccccc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H30FNO3/c1-19(2)30-24-12-9-7-11-23(24)27(20-14-16-21(29)17-15-20)25(30)13-8-6-10-22(31)18-26(32)33-28(3,4)5/h6-17,19H,18H2,1-5H3/b10-6+,13-8+
Formula Weight 447.221
Chemical Name or Material Fluvastatin 3-Oxo-4,6-diene O-tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.