missing translation for 'onlineSavingsMsg'
Learn More

τ-Fluvalinate (Mixture of 2 Diastereomers), TRC

Product Code. 30392842
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392842 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392842 Supplier Toronto Research Chemicals Supplier No. TRCF60110025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tau-Fluvalinate
CAS 102851-06-9
Molecular Formula C26 H22 Cl F3 N2 O3
Quantity 25 mg
Synonym Apistan, Fluvarol, Fluwarol, Klartan, Mavrik, Mavrik 25EC, Mavrik 2E, Mavrik Aquaflow, Mavrik EW, Minadox, Spur, Spur 22EW, Tau-fluvalinate, τ-Fluvalinate, N-[2-chloro-4-(trifluoromethyl)phenyl]-cyano(3-phenoxyphenyl)methyl D-Valine ester
InChI Formula InChI=1S/C26H22ClF3N2O3/c1-16(2)24(32-22-12-11-18(14-21(22)27)26(28,29)30)25(33)35-23(15-31)17-7-6-10-20(13-17)34-19-8-4-3-5-9-19/h3-14,16,23-24,32H,1-2H3/t23?,24-/m1/s1
SMILES CC(C)[C@@H](Nc1ccc(cc1Cl)C(F)(F)F)C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2
IUPAC Name [cyano-(3-phenoxyphenyl)methyl] (2R)-2-[2-chloro-4-(trifluoromethyl)anilino]-3-methylbutanoate
Molecular Weight (g/mol) 502.91
Formula Weight 502.1271
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.