missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Fluticasone propionate
GREENER_CHOICE

Product Code. 18772584
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18772584 10 mg 10mg
18772674 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 18772584

Brand: Tocris Bioscience™ 2007/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective high affinity glucocorticoid receptor agonist

High affinity, selective glucocorticoid receptor agonist (Kd = 0.5 nM). Potently stimulates glucocorticoid receptor-mediated transactivation of gene expression and enhances human eosinophil apoptosis (EC50 = 3.7 nM) in vitro. Inhibits mast cell accumulation in nasal mucosa following topical administration. Lipophilic anti-inflammatory agent with low oral bioavailability. Also potentiates KV1 channels (EC50 = 37 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S

Specifications

Quantity 10 mg
Formula Weight 500.57
Purity >98%
Molecular Weight (g/mol) 505.604
Chemical Name or Material Fluticasone propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.