missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone Propionate, Mikromol™

Product Code. 17922936
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
17922936 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 17922936

Brand: Mikromol MM1244.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fluticasone Propionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluticasone Propionate
CAS 80474-14-2
Analyte or Component Names Fluticasone Propionate
Shipping Condition Room Temperature
Molecular Formula C25 H31 F3 O5 S
Certifications/Compliance ISO 17034
Synonym Fluticasone propionate, 6alpha,9-Difluoro-17-[[(fluoromethyl)sulfanyl]carbonyl]-11beta-hydroxy-16alpha-methyl-3-oxoandrosta-1,4-dien-17alpha-yl propanoate, CCI 18781, Compound A (fluticasone propionate related), CCI 18781, unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X)
InChI Formula InChI=1S/C25H31F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1
SMILES CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF
IUPAC Name [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 500.57
Quantity 250 mg
Formula Weight 500.1844 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.