missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone (propionate), MedChemExpress

Product Code. 16454257
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16454257 50 mg 50mg
16438368 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16454257

Brand: Medchem Express HYB0154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fluticasone propionate, a potent topical anti-inflammatory corticosteroid, is a selective glucocorticoid receptor agonist, with an absolute affinity (KD) of 0.5 nM. Fluticasone propionate shows little or no activity at other steroid receptors. Anti-viral activity.

Fluticasone propionate, a potent topical anti-inflammatory corticosteroid, is a selective glucocorticoid receptor agonist, with an absolute affinity (KD) of 0.5 nM. Fluticasone propionate shows little or no activity at other steroid receptors. Anti-viral activity.

Specifications

Chemical Name or Material Fluticasone (propionate)
Physical Form Solid
CAS 80474-14-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C25H31F3O5S
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 100 mg/mL (199.77 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.00 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(CC)O[C@@]([C@]([C@@]1([H])C2)(C[C@H](O)[C@](F)([C@]3(C=C4)C)[C@@]1([H])C[C@H](F)C3=CC4=O)C)([C@@H]2C)C(SCF)=O
Molecular Weight (g/mol) 500.57
Formula Weight 500.57
Percent Purity 99.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.