missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluticasone Propionate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Fluticasone Propionate |
| CAS | 80474-14-2 |
| Analyte or Component Names | Fluticasone Propionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H31 F3 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fluticasone propionate, 6alpha,9-Difluoro-17-[[(fluoromethyl)sulfanyl]carbonyl]-11beta-hydroxy-16alpha-methyl-3-oxoandrosta-1,4-dien-17alpha-yl propanoate, CCI 18781, Compound A (fluticasone propionate related), CCI 18781, unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X) |
| InChI Formula | InChI=1S/C25H31F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1 |
| Solution Type | Neat |
| SMILES | CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?