missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluticasone Dimer Impurity (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | Fluticasone Dimer Impurity |
| CAS | 220589-37-7 |
| Molecular Formula | C43 H51 F5 O8 S |
| Quantity | 10 mg |
| Synonym | Androsta-1,4-diene-17-carboxylic acid, 6,9-difluoro-11,17-dihydroxy-16-methyl-3-oxo-, (6alpha,11beta,16alpha,17alpha)-6,9-difluoro-17-[[(fluoromethyl)thio]carbonyl]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-dien-17-yl ester, (6alpha,11beta,16alpha,17alpha)-, 6alpha,9-Difluoro-17-[[(fluoromethyl)sulfanyl]carbonyl]-11beta-hydroxy-16alpha-methyl-3-oxoandrosta-1,4-dien-17alpha-yl 6alpha,9-Difluoro-11beta,17-dihydroxy-16alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylate, Fluticasone Propionate Imp. G (EP), GR 112801X, BP Fluticasone Propionate Impurity G, Ph Eur Fluticasone Propionate Impurity G, USP Fluticasone Propionate Related Compound E, CAF (GW694301X), GW 872664X, TAF (GW675713X), 6-des-fluoro 698 (GW836850X), GSK465433A and GSK1322631A, 5-chloro-furoyl 698 (GSK180878A), TA-TAF dimer (GW871519X), Compound A (fluticasone propionate related), unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), GR 112801X, dithio (GR 269949X) |
| InChI Formula | InChI=1S/C43H51F5O8S/c1-20-11-24-26-15-30(45)28-13-22(49)7-9-36(28,3)40(26,47)32(51)17-38(24,5)42(20,55)34(53)56-43(35(54)57-19-44)21(2)12-25-27-16-31(46)29-14-23(50)8-10-37(29,4)41(27,48)33(52)18-39(25,43)6/h7-10,13-14,20-21,24-27,30-33,51-52,55H,11-12,15-19H2,1-6H3/t20-,21-,24+,25+,26+,27+,30+,31+,32+,33+,36+,37+,38+,39+,40+,41+,42+,43+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)O[C@@]5([C@H](C)C[C@H]6[C@@H]7C[C@H](F)C8=CC(=O)C=C[C@]8(C)[C@@]7(F)[C@@H](O)C[C@]56C)C(=O)SCF |
| IUPAC Name | [(6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylate |
| Molecular Weight (g/mol) | 822.92 |
| Formula Weight | 822.3225 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?