missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluticasone 17Beta-Carboxylic Acid Propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Fluticasone 17beta-Carboxylic Acid Propionate |
| CAS | 65429-42-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H30 F2 O6 |
| Quantity | 10 mg |
| Synonym | Androsta-1,4-diene-17-carboxylic acid, 6,9-difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)-, (6α,11β,16α,17α)-, 6α,9-difluoro-11β-hydroxy-16α-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17β-carboxylic acid, Fluticasone Propionate Imp. A (EP) |
| InChI Formula | InChI=1S/C24H30F2O6/c1-5-19(29)32-24(20(30)31)12(2)8-14-15-10-17(25)16-9-13(27)6-7-21(16,3)23(15,26)18(28)11-22(14,24)4/h6-7,9,12,14-15,17-18,28H,5,8,10-11H2,1-4H3,(H,30,31)/t12-,14+,15+,17+,18+,21+,22+,23+,24+/m1/s1 |
| SMILES | CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)O |
| IUPAC Name | (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid |
| Molecular Weight (g/mol) | 452.49 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?