missing translation for 'onlineSavingsMsg'
Learn More

Fluticasone 17-(((Fluoromethyl)thio)carbonyl) Furan-2-carboxylic Acid 11-Oxo, TRC

Product Code. 30401790
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401790 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401790 Supplier Toronto Research Chemicals Supplier No. TRCF59946910MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluticasone 17-(((Fluoromethyl)thio)carbonyl) Furan-2-carboxylic Acid 11-Oxo
CAS 397864-56-1
Molecular Formula C27 H27 F3 O6 S
Quantity 10 mg
Synonym (6S,8S,9R,10S,13S,14S,16R,17R)-6,9-Difluoro-17-(((fluoromethyl)thio)carbonyl)-10,13,16-trimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl furan-2-carboxylate
InChI Formula InChI=1S/C27H27F3O6S/c1-14-9-16-17-11-19(29)18-10-15(31)6-7-24(18,2)26(17,30)21(32)12-25(16,3)27(14,23(34)37-13-28)36-22(33)20-5-4-8-35-20/h4-8,10,14,16-17,19H,9,11-13H2,1-3H3/t14-,16+,17+,19+,24+,25+,26+,27+/m1/s1
SMILES C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)SCF
IUPAC Name [(6S,8S,9R,10S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
Molecular Weight (g/mol) 536.56
Formula Weight 536.148
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.