missing translation for 'onlineSavingsMsg'
Learn More

Flutamide, TRC

Product Code. 30391289
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391289 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391289 Supplier Toronto Research Chemicals Supplier No. TRCF59885050MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flutamide
CAS 13311-84-7
Molecular Formula C11 H11 F3 N2 O3
Quantity 50 mg
Synonym Flutamide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide, α,α,α-Trifluoro-2-methyl-4'-nitro-m-propionotoluidide, 4'-Nitro-3'-trifluoromethylisobutyranilide, 4-Nitro-3-(trifluoromethyl)isobutyranilide, Euflex, Eulexin, Flucinom, Flutamide, N-(Isopropylcarbonyl)-4-nitro-3-trifluoromethylaniline, NSC 147834, NSC 215876, Niftholide, Niftolide, Odyne, Sch 13521
InChI Formula InChI=1S/C11H11F3N2O3/c1-6(2)10(17)15-7-3-4-9(16(18)19)8(5-7)11(12,13)14/h3-6H,1-2H3,(H,15,17)
SMILES CC(C)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 276.21
Formula Weight 276.0722
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.