missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Flutamide
GREENER_CHOICE

Product Code. 18732634
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732634 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732634 Supplier Tocris Bioscience™ Supplier No. 4094/50
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Non-steroidal androgen receptor antagonist

Flutamide is a non-steroidal antiandrogen; competitive antagonist of the androgen receptor. Blocks the biological activity of testosterone in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 13311-84-7
Quantity 50 mg
Molecular Formula C11H11F3N2O3
Formula Weight 276.21
Purity 98%
Molecular Weight (g/mol) 276.22
Chemical Name or Material Flutamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.